Assay Details
| Gene Symbol: LOC100387916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758966.3 | XP_002759012.1 | 6 - 7 | 1451 | 82 | |
| XM_008982729.2 | XP_008980977.1 | 7 - 8 | 987 | 82 | ||
| XM_008982730.2 | XP_008980978.1 | 6 - 7 | 642 | 82 | ||
| XM_017965376.1 | XP_017820865.1 | 7 - 8 | 701 | 82 | ||
| XM_017965377.1 | XP_017820866.1 | 6 - 7 | 630 | 82 | ||
| XM_017965378.1 | XP_017820867.1 | 6 - 7 | 692 | 82 | ||
| XM_017965379.1 | XP_017820868.1 | 3 - 4 | 249 | 82 | ||
More Information
Set Membership: |
|

LOC100387916
Genomic Map