Assay Details
| Gene Symbol: LOC100387916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758966.3 | XP_002759012.1 | 7 - 8 | 1499 | 88 | |
| XM_008982729.2 | XP_008980977.1 | 8 - 9 | 1035 | 88 | ||
| XM_008982730.2 | XP_008980978.1 | 7 - 8 | 690 | 88 | ||
| XM_017965376.1 | XP_017820865.1 | 8 - 9 | 749 | 88 | ||
| XM_017965377.1 | XP_017820866.1 | 7 - 8 | 678 | 88 | ||
| XM_017965378.1 | XP_017820867.1 | 7 - 8 | 740 | 88 | ||
| XM_017965379.1 | XP_017820868.1 | 4 - 5 | 297 | 88 | ||
More Information
Set Membership: |
|

LOC100387916
Genomic Map