Assay Details
| Gene Symbol: FAM168B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749222.3 | XP_002749268.1 | 3 - 4 | 184 | 94 | |
| XM_008998538.2 | XP_008996786.1 | 3 - 4 | 419 | 94 | ||
| XM_008998539.2 | XP_008996787.1 | 3 - 4 | 236 | 94 | ||
More Information
Set Membership: |
|

FAM168B
Genomic Map