Assay Details
| Gene Symbol: FRMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734306.3 | XP_003734354.1 | 10 - 11 | 1460 | 58 | |
| XM_009008473.2 | XP_009006721.1 | 10 - 11 | 1543 | 58 | ||
More Information
Set Membership: |
|

FRMD8
Genomic Map