Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749566.3 | XP_002749612.1 | 7 - 8 | 1639 | 85 | |
| XM_003733204.3 | XP_003733252.2 | 7 - 8 | 1882 | 85 | ||
| XM_008999098.2 | XP_008997346.1 | 7 - 8 | 1885 | 85 | ||
| XM_008999099.2 | XP_008997347.1 | 7 - 8 | 1478 | 85 | ||
More Information
Set Membership: |
|

NAB1
Genomic Map