Assay Details
| Gene Symbol: AURKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747695.4 | XP_002747741.1 | 3 - 4 | 498 | 71 | |
| XM_008996154.2 | XP_008994402.1 | 3 - 4 | 484 | 71 | ||
| XM_008996155.2 | XP_008994403.1 | 3 - 4 | 342 | 71 | ||
More Information
Set Membership: |
|

AURKA
Genomic Map