Assay Details
| Gene Symbol: SAP30BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748739.4 | XP_002748785.1 | 8 - 9 | 852 | 74 | |
| XM_008997754.1 | XP_008996002.1 | 8 - 9 | 738 | 74 | ||
| XM_008997755.2 | XP_008996003.1 | 7 - 8 | 617 | 74 | ||
| XM_008997756.2 | XP_008996004.1 | 7 - 8 | 617 | 74 | ||
More Information
Set Membership: |
|

SAP30BP
Genomic Map