Assay Details
| Gene Symbol: EBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762835.4 | XP_002762881.1 | 4 - 5 | 2064 | 69 | |
| XM_008989231.2 | XP_008987479.1 | 5 - 6 | 2300 | 69 | ||
| XM_008989232.2 | XP_008987480.1 | 4 - 5 | 629 | 69 | ||
| XM_008989233.2 | XP_008987481.1 | 5 - 6 | 2253 | 69 | ||
| XM_008989234.2 | XP_008987482.1 | 4 - 5 | 2017 | 69 | ||
| XM_017968574.1 | XP_017824063.1 | 4 - 5 | 576 | 69 | ||
More Information
Set Membership: |
|

EBP
Genomic Map