Assay Details
| Gene Symbol: BICRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968084.1 | XP_017823573.1 | 6 - 7 | 2637 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BICRA| Gene Symbol: BICRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968084.1 | XP_017823573.1 | 6 - 7 | 2637 | 106 | |
Set Membership: |
|