Assay Details
| Gene Symbol: GORASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749360.3 | XP_002749406.1 | 7 - 8 | 1610 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GORASP2| Gene Symbol: GORASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749360.3 | XP_002749406.1 | 7 - 8 | 1610 | 64 | |
Set Membership: |
|