Assay Details
| Gene Symbol: ACADVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996621.2 | XP_008994869.1 | 12 - 13 | 1332 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACADVL| Gene Symbol: ACADVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996621.2 | XP_008994869.1 | 12 - 13 | 1332 | 58 | |
Set Membership: |
|