Assay Details
| Gene Symbol: TWF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752353.4 | XP_002752399.3 | 2 - 3 | 212 | 83 | |
| XM_003733725.2 | XP_003733773.1 | 2 - 3 | 212 | 83 | ||
More Information
Set Membership: |
|

TWF1
Genomic Map