Assay Details
| Gene Symbol: PSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009765.1 | XP_009008013.1 | 14 - 15 | 1701 | 59 | |
| XM_009009766.1 | XP_009008014.1 | 14 - 15 | 1698 | 59 | ||
| XM_009009767.1 | XP_009008015.1 | 13 - 14 | 1692 | 59 | ||
More Information
Set Membership: |
|

PSAP
Genomic Map