Assay Details
| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966629.1 | XP_017822118.1 | 17 - 18 | 2857 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CR1| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966629.1 | XP_017822118.1 | 17 - 18 | 2857 | 75 | |