Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743666.3 | XP_002743712.1 | 2 - 3 | 424 | 101 | |
| XM_009009058.2 | XP_009007306.1 | 3 - 4 | 467 | 101 | ||
More Information
Set Membership: |
|

LIF
Genomic Map