Assay Details
| Gene Symbol: STK26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763266.4 | XP_002763312.1 | 2 - 3 | 336 | 63 | |
| XM_017968612.1 | XP_017824101.1 | 2 - 3 | 337 | 63 | ||
More Information
Set Membership: |
|

STK26
Genomic Map