Assay Details
| Gene Symbol: LOC100409351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967413.1 | XP_017822902.1 | 8 - 9 | 1847 | 113 | |
| XM_017967414.1 | XP_017822903.1 | 8 - 9 | 1661 | 113 | ||
| XM_017967415.1 | XP_017822904.1 | 8 - 9 | 1847 | 113 | ||
| XM_017967416.1 | XP_017822905.1 | 8 - 9 | 1847 | 113 | ||
| XM_017967417.1 | XP_017822906.1 | 8 - 9 | 1720 | 113 | ||
More Information
Set Membership: |
|

LOC100409351
Genomic Map