Assay Details
| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762567.3 | XP_002762613.1 | 2 - 3 | 324 | 72 | |
| XM_002762568.3 | XP_002762614.1 | 2 - 3 | 344 | 72 | ||
| XM_008988853.2 | XP_008987101.1 | 2 - 3 | 344 | 72 | ||
| XM_008988854.1 | XP_008987102.1 | 2 - 3 | 510 | 72 | ||
| XM_008988855.2 | XP_008987103.1 | 2 - 3 | 326 | 72 | ||
More Information
Set Membership: |
|

CHMP2A
Genomic Map