Assay Details
| Gene Symbol: BDKRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006512.2 | XP_009004760.1 | 2 | 1572 | 100 | |
| XM_009006515.2 | XP_009004763.1 | 2 | 1081 | 100 | ||
More Information
Set Membership: |
|

BDKRB1
Genomic Map