Assay Details
| Gene Symbol: METTL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996349.2 | XP_008994597.1 | 7 - 8 | 952 | 75 | |
| XM_008996350.2 | XP_008994598.1 | 6 - 7 | 789 | 75 | ||
More Information
Set Membership: |
|

METTL16
Genomic Map