Assay Details
| Gene Symbol: ZNF567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987816.2 | XP_008986064.1 | 4 - 5 | 623 | 74 | |
| XM_008987817.2 | XP_008986065.1 | 4 - 5 | 381 | 74 | ||
| XM_017968126.1 | XP_017823615.1 | 5 - 6 | 749 | 74 | ||
| XM_017968127.1 | XP_017823616.1 | 5 - 6 | 484 | 74 | ||
| XM_017968128.1 | XP_017823617.1 | 5 - 6 | 642 | 74 | ||
| XM_017968129.1 | XP_017823618.1 | 5 - 6 | 736 | 74 | ||
More Information
Set Membership: |
|

ZNF567
Genomic Map