Assay Details
| Gene Symbol: CYB5R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746774.3 | XP_002746820.1 | 5 - 6 | 572 | 73 | |
| XM_008994727.2 | XP_008992975.1 | 5 - 6 | 574 | 73 | ||
| XM_008994728.2 | XP_008992976.1 | 5 - 6 | 574 | 73 | ||
More Information
Set Membership: |
|

CYB5R4
Genomic Map