Assay Details
| Gene Symbol: ALAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758411.4 | XP_002758457.1 | 10 - 11 | 1956 | 62 | |
| XM_008981771.2 | XP_008980019.1 | 10 - 11 | 1944 | 62 | ||
More Information
Set Membership: |
|

ALAS1
Genomic Map