Assay Details
| Gene Symbol: BLOC1S5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746239.3 | XP_002746285.2 | 1 - 2 | 480 | 82 | |
| XM_008993995.2 | XP_008992243.2 | 1 - 2 | 480 | 82 | ||
| XM_008993996.2 | XP_008992244.2 | 1 - 2 | 367 | 82 | ||
| XM_008993998.2 | XP_008992246.2 | 1 - 2 | 367 | 82 | ||
| XM_017970921.1 | XP_017826410.1 | 1 - 2 | 367 | 82 | ||
| XM_017970922.1 | XP_017826411.1 | 1 - 2 | 367 | 82 | ||
More Information
Set Membership: |
|

BLOC1S5
Genomic Map