Assay Details
| Gene Symbol: POLI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757247.3 | XP_002757293.1 | 4 - 5 | 677 | 89 | |
| XM_008979840.2 | XP_008978088.1 | 4 - 5 | 537 | 89 | ||
| XM_008979841.2 | XP_008978089.1 | 4 - 5 | 609 | 89 | ||
| XM_017963479.1 | XP_017818968.1 | 2 - 3 | 411 | 89 | ||
More Information
Set Membership: |
|

POLI
Genomic Map