Assay Details
| Gene Symbol: TOGARAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005986.2 | XP_009004234.1 | 11 - 12 | 4050 | 102 | |
| XM_009005987.2 | XP_009004235.1 | 11 - 12 | 4052 | 102 | ||
| XM_017977072.1 | XP_017832561.1 | 10 - 11 | 3744 | 102 | ||
| XM_017977073.1 | XP_017832562.1 | 11 - 12 | 4057 | 102 | ||
More Information
Set Membership: |
|

TOGARAM1
Genomic Map