Assay Details
| Gene Symbol: IVD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753548.3 | XP_002753594.1 | 7 - 8 | 899 | 71 | |
| XM_009005559.1 | XP_009003807.1 | 7 - 8 | 899 | 71 | ||
| XM_017976926.1 | XP_017832415.1 | 7 - 8 | 899 | 71 | ||
More Information
Set Membership: |
|

IVD
Genomic Map