Assay Details
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003735165.3 | XP_003735213.1 | 1 - 2 | 150 | 83 | |
XM_008984785.2 | XP_008983033.1 | 1 - 2 | 149 | 86 | ||
XM_008984786.2 | XP_008983034.1 | 1 - 2 | 160 | 86 | ||
XM_008984787.2 | XP_008983035.1 | 1 - 2 | 134 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
