Assay Details
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003735165.3 | XP_003735213.1 | 5 - 6 | 807 | 63 | |
XM_008984785.2 | XP_008983033.1 | 5 - 6 | 809 | 63 | ||
XM_008984786.2 | XP_008983034.1 | 5 - 6 | 820 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
