Assay Details
| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806661.4 | XP_002806707.2 | 3 - 4 | 1242 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPFFR2| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806661.4 | XP_002806707.2 | 3 - 4 | 1242 | 95 | |
Set Membership: |
|