Assay Details
| Gene Symbol: GPD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983957.2 | XP_008982205.1 | 7 - 8 | 1161 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPD1L| Gene Symbol: GPD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983957.2 | XP_008982205.1 | 7 - 8 | 1161 | 72 | |
Set Membership: |
|