Assay Details
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744988.4 | XP_002745034.2 | 6 - 7 | 758 | 91 | |
XM_008992080.2 | XP_008990328.1 | 7 - 8 | 836 | 91 | ||
XM_017969929.1 | XP_017825418.1 | 5 - 6 | 546 | 91 | ||
GenBank mRNA | EF214926.1 | - | 1 - 2 | 124 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
