Assay Details
| Gene Symbol: ATP6V0C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755788.4 | XP_002755834.1 | 2 - 3 | 643 | 76 | |
| GenBank mRNA | EF215006.1 | - | 1 - 2 | 47 | 76 | |
More Information
Set Membership: |
|

ATP6V0C
Genomic Map