Assay Details
Gene Symbol: IRAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008982141.2 | XP_008980389.1 | 1 - 2 | 172 | 59 | |
XM_008982142.2 | XP_008980390.1 | 1 - 2 | 172 | 59 | ||
XM_008982143.2 | XP_008980391.1 | 1 - 2 | 172 | 59 | ||
XM_008982144.2 | XP_008980392.1 | 1 - 2 | 172 | 59 | ||
XM_008982145.2 | XP_008980393.1 | 1 - 2 | 172 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
