Assay Details
| Gene Symbol: OSBPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747738.3 | XP_002747784.1 | 10 - 11 | 1219 | 67 | |
| XM_002747739.3 | XP_002747785.1 | 10 - 11 | 1204 | 67 | ||
| XM_008996227.2 | XP_008994475.1 | 10 - 11 | 1240 | 67 | ||
| XM_008996228.1 | XP_008994476.1 | 10 - 11 | 1240 | 67 | ||
More Information
Set Membership: |
|

OSBPL2
Genomic Map