Assay Details
| Gene Symbol: KCTD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756047.4 | XP_002756093.1 | 5 - 6 | 966 | 97 | |
| XM_017978853.1 | XP_017834342.1 | 6 - 7 | 1029 | 97 | ||
More Information
Set Membership: |
|

KCTD13
Genomic Map