Assay Details
| Gene Symbol: MRPS25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734896.3 | XP_003734944.1 | 2 - 3 | 378 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPS25| Gene Symbol: MRPS25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734896.3 | XP_003734944.1 | 2 - 3 | 378 | 77 | |
Set Membership: |
|