Assay Details
| Gene Symbol: GUCY1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745302.3 | XP_002745348.1 | 3 - 4 | 631 | 96 | |
| XM_002745303.4 | XP_002745349.1 | 3 - 4 | 692 | 96 | ||
| XM_008992555.2 | XP_008990803.1 | 3 - 4 | 681 | 96 | ||
| XM_008992556.2 | XP_008990804.1 | 3 - 4 | 753 | 96 | ||
| XM_008992557.2 | XP_008990805.1 | 2 - 3 | 625 | 96 | ||
| XM_008992559.2 | XP_008990807.1 | 4 - 5 | 668 | 96 | ||
| XM_017970181.1 | XP_017825670.1 | 2 - 3 | 1668 | 96 | ||
| XM_017970182.1 | XP_017825671.1 | 3 - 4 | 692 | 96 | ||
| XM_017970183.1 | XP_017825672.1 | 4 - 5 | 718 | 96 | ||
| XM_017970184.1 | XP_017825673.1 | 3 - 4 | 540 | 96 | ||
| XM_017970185.1 | XP_017825674.1 | 4 - 5 | 718 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008992559.2, XM_017970183.1, XM_017970184.1, XM_017970185.1 transcripts.
Set Membership: |
|

GUCY1A1
Genomic Map