Assay Details
| Gene Symbol: SLC31A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001460.2 | XP_008999708.1 | 2 - 3 | 359 | 66 | |
| XM_017975639.1 | XP_017831128.1 | 2 - 3 | 323 | 66 | ||
More Information
Set Membership: |
|

SLC31A2
Genomic Map