Assay Details
| Gene Symbol: PDE7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965370.1 | XP_017820859.1 | 3 - 4 | 285 | 98 | |
| XM_017965371.1 | XP_017820860.1 | 3 - 4 | 1643 | 98 | ||
| XM_017965372.1 | XP_017820861.1 | 3 - 4 | 3898 | 98 | ||
More Information
Set Membership: |
|

PDE7A
Genomic Map