Assay Details
| Gene Symbol: METAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745570.4 | XP_002745616.1 | 8 - 9 | 926 | 67 | |
| XM_017970405.1 | XP_017825894.1 | 8 - 9 | 926 | 67 | ||
More Information
Set Membership: |
|

METAP1
Genomic Map