Assay Details
| Gene Symbol: GPNMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733579.3 | XP_003733627.1 | 1 - 2 | 361 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPNMB| Gene Symbol: GPNMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733579.3 | XP_003733627.1 | 1 - 2 | 361 | 73 | |
Set Membership: |
|