Assay Details
| Gene Symbol: EIF1AD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755612.4 | XP_002755658.1 | 5 - 6 | 610 | 62 | |
| XM_009008502.2 | XP_009006750.1 | 5 - 6 | 670 | 62 | ||
| XM_017978289.1 | XP_017833778.1 | 5 - 6 | 568 | 62 | ||
More Information
Set Membership: |
|

EIF1AD
Genomic Map