Assay Details
| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755792.3 | XP_002755838.1 | 2 - 3 | 190 | 99 | |
| XM_009008931.2 | XP_009007179.1 | 2 - 3 | 190 | 99 | ||
| XM_017978625.1 | XP_017834114.1 | 2 - 3 | 190 | 99 | ||
| XM_017978626.1 | XP_017834115.1 | 2 - 3 | 190 | 99 | ||
| XM_017978627.1 | XP_017834116.1 | 2 - 3 | 190 | 99 | ||
| XM_017978628.1 | XP_017834117.1 | 2 - 3 | 190 | 99 | ||
More Information
Set Membership: |
|

TEDC2
Genomic Map