Assay Details
| Gene Symbol: LOC100392818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752489.3 | XP_002752535.1 | 4 - 5 | 1337 | 74 | |
| XM_009003833.2 | XP_009002081.1 | 4 - 5 | 1637 | 74 | ||
| XM_009003834.2 | XP_009002082.1 | 5 - 6 | 1787 | 74 | ||
| XM_009003836.2 | XP_009002084.1 | 5 - 6 | 1488 | 74 | ||
More Information
Set Membership: |
|

LOC100392818
Genomic Map