Assay Details
| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996277.2 | XP_008994525.1 | 4 - 5 | 1032 | 72 | |
| XM_017972172.1 | XP_017827661.1 | 4 - 5 | 1035 | 72 | ||
| XM_017972173.1 | XP_017827662.1 | 4 - 5 | 1035 | 72 | ||
More Information
Set Membership: |
|

SLC2A4RG
Genomic Map