Assay Details
| Gene Symbol: PNLIPRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756620.2 | XP_002756666.2 | 7 - 8 | 828 | 81 | |
| XM_009010336.1 | XP_009008584.1 | 7 - 8 | 828 | 81 | ||
More Information
Set Membership: |
|

PNLIPRP2
Genomic Map