Assay Details
| Gene Symbol: MMP23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750187.3 | XP_002750233.1 | 1 - 2 | 726 | 92 | |
| XM_009000251.2 | XP_008998499.1 | 1 - 2 | 728 | 92 | ||
| XM_009000252.2 | XP_008998500.1 | 1 - 2 | 728 | 92 | ||
| XM_009000253.2 | XP_008998501.1 | 1 - 2 | 728 | 92 | ||
More Information
Set Membership: |
|

MMP23B
Genomic Map