Assay Details
| Gene Symbol: MMP23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750187.3 | XP_002750233.1 | 2 - 3 | 853 | 54 | |
| XM_009000251.2 | XP_008998499.1 | 2 - 3 | 855 | 54 | ||
| XM_009000252.2 | XP_008998500.1 | 2 - 3 | 855 | 54 | ||
| XM_009000253.2 | XP_008998501.1 | 2 - 3 | 855 | 54 | ||
More Information
Set Membership: |
|

MMP23B
Genomic Map