Assay Details
| Gene Symbol: TMEM79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760098.3 | XP_002760144.1 | 2 - 3 | 1613 | 84 | |
| XM_017966353.1 | XP_017821842.1 | 1 - 2 | 988 | 84 | ||
| XM_017966354.1 | XP_017821843.1 | 2 - 3 | 1762 | 84 | ||
More Information
Set Membership: |
|

TMEM79
Genomic Map